Learn More
Masterflex™ C-Flex™ L/S™ High Performance Precision Pump Tubing
Product Code. 11720999
Click to view available options
Tubing Size:
15
24
35
36
Unit Size:
Each
Product Code. 11720999
Brand: Masterflex™ 0642436
Only null left
Add to basket
Add to basket
Ensure optimal Masterflex™ pump performance with this tubing. Masterflex™ C-Flex™ L/S™ High Performance Precision Pump Tubing combines biocompatibility of silicone with chemical resistance similar to Tygon™. Ideal for applications involving pressure, suction lift and viscous fluids.
C-Flex™ tubing features exceptional tensile and tear strength, good biocompatibility, low gas permeability and a smooth surface. General purpose lab tubing for applications requiring high performance and long life.
  • Thicker wall compared to Precision pump tubing
  • Lot-to-lot consistency for superior accuracy and repeatability
  • Provides longer life than other platinum and peroxide silicone tubing formulations
  • Very low protein binding
  • Heat sealable, weldable
  • Better resistance to pH fluctuation
  • Complies with USP Class VI, EP, REACH, RoHS, ADCF
  • Economical
  • Opaque White
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 9.7 mm
Material C-Flex
Overall Length (Metric) 760 cm
For Use With (Application) For general-purpose lab uses requiring high performance and long life
Min. Operating Temperature (Metric) -40°C
Tubing Size 36
Flow Rate 4.8 to 2900mL/min.
Hose Barb 3/8 in.
Height (Metric) 8.2 m
Gas Permeability Low
Color Opaque White
Maximum Pressure 20 psi
Vacuum Level 610 mm
Max. Operating Temperature (Metric) 135°C
Product Type Pump Tubing