Learn More
Saint Gobain Life Sciences C-Flex (ACDF) Tubing
Provides exceptional tensile and tear strength, good biocompatibility, and excellent chemical resistance. Saint Gobain Life Sciences C-Flex (ACDF) Tubing is transparent with low gas permeability and a smooth surface.
Brand: Saint Gobain Life Sciences 06432-59
This item is not returnable.
View return policy
Description
- C-Flex ADCF is animal-derived component-free.
- Exceptional tensile and tear strength with good biocompatibility
- Excellent chemical resistance
- Low gas permeability
- Smooth surface
- Transparent.
- Resin meets USP and Class VI requirements. Tubing is manufactured according to GMP.
- Temperature range: 45 to 135°C (50 to 275°F)
- Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Specifications
| USP, USDA, GMP | |
| 9.6 mm | |
| 15.9 mm | |
| 3.2 mm | |
| 1 Pk. | |
| Transparent | |
| 135°C |
| Autoclavable | |
| C-Flex | |
| 15.24 m | |
| -45°C | |
| Sterile | |
| 23 psi |