missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Abl Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2755.00 NOK - 4135.00 NOK
Specifications
| Antigen | c-Abl |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452862
|
Novus Biologicals
NBP1-90288-25ul |
25 μL |
2755.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18436591
|
Novus Biologicals
NBP1-90288 |
0.1 mL |
4135.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Abl Polyclonal antibody specifically detects c-Abl in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| c-Abl | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Signal Transduction, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol | |
| 25 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RVRRHKHSSESPGRDKGKLSRLKPAPPPPPAASAGKAGGKPSQSPSQEAAGEAVLGAKTKATSLVDAVNSDAAK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title