Learn More
Cytiva C 10/10 Column
Product Code. 10221110 Shop All Cytiva Products
Click to view available options
Diameter (Metric):
10 mm
Unit Size:
Each
Product Code. 10221110
Brand: Cytiva 19503501
Only null left
Add to basket
Add to basket
Economical empty columns suitable for low pressure liquid chromatography, and gravity feed at lower volumes typically used at research scale.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 10 mm
For Use With Low Pressure Liquid Chromatography
Includes Polyethylene Tubing (1 mm I.D.), Bed Support Screens, Snap-On Net Rings, and Instructions
Diameter (Metric) Inner 10 mm
Diameter (Metric) 10 mm