missing translation for 'onlineSavingsMsg'
Learn More

BZW1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19917-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889384

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

Enhances histone H4 gene transcription but does not seem to bind DNA directly.
Specifications

Specifications

BZW1
Polyclonal
Unconjugated
BZW1
BZAP45, BZW1, KIAA0005, Nbla10236, Protein Orf
Rabbit
Antigen Affinity Chromatography
RUO
363232, 66882, 9689
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P7P5, Q7L1Q6, Q9CQC6
Bzw1
BZW1 Fusion Protein Ag13830
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers