Learn More
BZW1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19917-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Enhances histone H4 gene transcription but does not seem to bind DNA directly.Specifications
BZW1 | |
Polyclonal | |
Unconjugated | |
BZW1 | |
BZAP45, BZW1, KIAA0005, Nbla10236, Protein Orf | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363232, 66882, 9689 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P7P5, Q7L1Q6, Q9CQC6 | |
Bzw1 | |
BZW1 Fusion Protein Ag13830 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |