missing translation for 'onlineSavingsMsg'
Learn More

BX795, MedChemExpress

Product Code. 16406737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406737

Brand: Medchem Express HY10514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BX795 is a potent and selective inhibitor of PDK1, with an IC50 of 6 nM. BX795 is also a potent and relatively specific inhibitor of TBK1 and IKKɛ, with an IC50 of 6 and 41 nM, respectively. BX795 blocks phosphorylation of S6K1, Akt, PKCδ, and GSK3β, and has lower selectivity over PKA, PKC, c-Kit, GSK3β etc. BX795 modulates autophagy.

BX795 is a potent and selective inhibitor of PDK1, with an IC50 of 6 nM. BX795 is also a potent and relatively specific inhibitor of TBK1 and IKKɛ, with an IC50 of 6 and 41 nM, respectively. BX795 blocks phosphorylation of S6K1, Akt, PKCδ, and GSK3β, and has lower selectivity over PKA, PKC, c-Kit, GSK3β etc. BX795 modulates autophagy.

Specifications

Chemical Name or Material BX795
Physical Form Solid
CAS 702675-74-9
Color Light Gray
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H26IN7O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 33.33 mg/mL (56.35 mM; Need ultrasonic)
SMILES O=C(NCCCNC1=C(C=NC(NC2=CC=CC(NC(N3CCCC3)=O)=C2)=N1)I)C4=CC=CS4
Molecular Weight (g/mol) 591.47
Formula Weight 591.47
Percent Purity 98.93%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.