Learn More
BX471 hydrochloride, MedChemExpress
Product Code. 16474978
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16474978 10 mg 10mg
16441227 50 mg 50mg
2 options
Product Code. 16474978
Brand: Medchem Express HY12080A
MedChemExpress BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.
BX471 hydrochloride (ZK-811752 hydrochloride) is a potent, selective non-peptide CCR1 antagonist with Ki of 1 nM for human CCR1, and exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4.
Specifications
Chemical Name or Material BX471 hydrochloride
CAS 288262-96-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25Cl2FN4O3
Quantity 10 mg
Synonym ZK-811752 hydrochloride
Solubility Information DMSO : 150 mg/mL (318.23 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(N)NC1=CC(Cl)=CC=C1OCC(N2[C@H](C)CN(CC3=CC=C(F)C=C3)CC2)=O.[H]Cl
Molecular Weight (g/mol) 471.35
Formula Weight 471.35
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation