Learn More
BRANDTECH™ VACUUBRAND™ BVC Fluid Aspiration Systems for Cell Culture
GREENER_CHOICE
Product Code. 12683315
Change view
Click to view available options
Overall Height (Metric):
43 cm
49 cm
50 cm
Overall Length (Metric):
23 cm
25 cm
40.8 cm
Model:
BVC Basic
BVC Basic G
BVC Control
BVC Control G
BVC Professional
BVC Professional G
Unit Size:
Each
Plug Type:
CEE
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Height (Metric) Overall Length (Metric) Model Unit Size Plug Type
12683315 50 cm 40.8 cm BVC Control Each -
12653315 49 cm 25 cm BVC Basic Each -
12663315 43 cm 40.8 cm BVC Control G Each CEE
12603325 43 cm 40.8 cm BVC Professional G Each CEE
12693315 50 cm 40.8 cm BVC Professional Each CEE
12673315 43 cm 23 cm BVC Basic G Each -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12683315 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20727200
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 14-12-2026
BVC Fluid Aspiration Systems for Cell Culture are designed for optimal vacuum control, efficient supernatant aspiration, and safe disposal of biological fluids.
Basic, Control and Professional models to serve your fluid aspiration needs.
  • 2 or 4 L vessels for safe collection and disposal of your biological waste
  • Autoclavable hydrophobic 0.2 μm biofilter protects pump or other vacuum supply from biohazardous aerosols
  • The VacuuHandControl (VHC) keeps all fluid within aspiration tubing for the ultimate in containment and cleaning convenience
  • G models feature a 2 L glass collection vessel with scratch and fragment protection coating and true bleach resistance
  • Standard versions include a 4 L heavy duty polypropylene vacuum bottle for increased capacity
  • Control and Professional models feature powerful VACUUBRAND™ chemistry design vacuum pumps with exceptionally long service life and enough power for two users
  • Streamlined design incorporates bottle, controls and pump (Control and Professional models), in a lab friendly small footprint
  • Quick connectors are supplied with the standard (non-G) Professional, but can be added to the other five models
BVC Basic
  • The unpowered version, suitable for use with a VACUU LAN vacuum network, central vacuum system or shared pump
  • Provides a suitable vacuum level in the bottle for aspiration, and shuts off connection to the line when appropriate vacuum level is reached
  • Supplied with a stable and easy-to-clean powder coated metal stand, 0.2 μm aerosol filter, and VacuuHandControl
  • Available in heavy duty polypropylene, or bleach resistant glass versions
BVC Control
  • The BVC Control features a powerful whisper-quiet VACUUBRAND chemistry-design vacuum pump, eliminating the need for central vacuum supply
  • Potentially biohazardous materials can be contained in a local area
  • The pump is turned on and off automatically on demand when using the VacuuHandControl which further reduces the noise level
  • An easy to clean touch panel adjusts and indicates the vacuum level switch point
  • Available with either a 4 L polypropylene, or 2 L bleach resistant glass bottle
BVC Professional
  • Includes an external liquid level sensor, which operates without contacting bottle contents
  • Special disinfection program allows the aspiration of disinfectant after the system is shut down to decontaminate the VacuuHandControl
  • The 4 L polypropylene bottle version is supplied with quick connectors as standard
  • The 2 L glass bottle 'bleach proof' version includes hose barb connections; optional quick connectors are available
Available Accessories:
  • Replacement filter 638266
  • Replacement VHCpro tubing (sold per meter, minimum order 3 m) 636156
  • VHCpro hand controller (one supplied with unit) 688061
  • VHCpro stand 635680
  • Pipette tip ejector for VHCpro 635638
  • 8-channel adapter with tip ejector for VHCpro 635679
  • BVC shuttle (wheeled cart to facilitate access to BVC when under a BSC) 696880
  • Quick connect for second user (between VHCpro and bottle) 635807
  • Standard connect for second user 699943
  • Quick connect (between bottle and pump) 635808
  • Replacement 4 L PP bottle (with cap) 635810
  • Replacement 2 L glass bottle (with cap) 635809
  • 2 L glass bottle adapter 635839
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 150 mbar (112 Torr)
Includes 4 L Polypropylene Bottle, Vacuum Pump, 0.2 μm Biofilter
Electrical Requirements 230 V 50/60 Hz
Model BVC Control
Noise Level 45 dB (A)
Weight (English) 16.09 lb.
Weight (Metric) 7.3 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 0.4/0.5 CFM
Frequency 50/60 Hz
Overall Height (English) 19.68 in.
Overall Height (Metric) 50 cm
Overall Length (English) 16.06 in.
Overall Length (Metric) 40.8 cm
Overall Width (English) 7.63 in.
Pump Type Chemistry Diaphragm Pump
Enclosure Rating IP40
Maximum Pressure 1.1 bar
Voltage 230 V
Storage Requirements -10°C to +60°C