Learn More
Butyric Anhydride, Sigma-Aldrich™
Product Code. 15878310
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878310 1 L 1 L
15868310 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15878310 Supplier Sigma Aldrich Supplier No. 8005031000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-31-0
Specifications
Melting Point -75°C
Boiling Point 195°C to 199°C/1013 hPa
Density 0.963 g/cm3 at 20°C
Quantity 1 L
Molecular Formula (CH3CH2CH2CO)2O
MDL Number MFCD00009389
Synonym Butyric anhydride, Butyric anhydride
Molecular Weight (g/mol) 158.19
Grade Ungraded
Chemical Name or Material Butyric Anhydride