Learn More
Butyric Acid, Sigma-Aldrich™
Product Code. 15856240
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
50 kg
Unit Size:
1 L
100mL
2.5 L
50kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856240 2.5 L 2.5 L
15828280 100 mL 100mL
15846240 1 L 1 L
15838280 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15856240 Supplier Sigma Aldrich Supplier No. 8004572500
 Restricted Item
Estimated Shipment: 25-01-2027
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-92-6
Specifications
Melting Point -8°C to 5°C
Boiling Point 162°C to 165°C/1013 mbar
Density 0.96 g/cm3 at 20°C
Quantity 2.5 L
MDL Number MFCD00002814
Synonym Butyric acid, Butyric acid
Molecular Weight (g/mol) 88.11
Grade Ungraded
Chemical Name or Material Butyric Acid