missing translation for 'onlineSavingsMsg'
Learn More

Butyltinhydroxide-oxide, Sigma-Aldrich™

Product Code. 15874701
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15874701 100 g 100g
15864701 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15874701 Supplier Sigma Aldrich Supplier No. 8401260100
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2273-43-0

Specifications

Boiling Point >350°C/1013 hPa
MDL Number MFCD00013927
Quantity 100 g
Synonym Butyltinhydroxide-oxide, Butylhydroxyoxo stannane, Butyltinhydroxide-oxide
Molecular Weight (g/mol) 208.83
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.