Learn More
Macherey-Nagel™ Butyl Phase Polygosil™ C4
Product Code. 10566291 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
10 g
100 g
Particle Size:
7 μm
Pore Size:
1000 Å
300 Å
Unit Size:
100g
10g
Product Code. 10566291
Brand: Macherey-Nagel™ 711680.100