Learn More
Butyl 4-Aminobenzoate(Butamben), TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Butamben |
| CAS | 94-25-7 |
| Molecular Formula | C11 H15 N O2 |
| Quantity | 50 g |
| Synonym | Butyl aminobenzoate, Benzoic acid, 4-amino-, butyl ester, Benzoic acid, p-amino-, butyl ester (6CI,7CI,8CI), 4-(Butoxycarbonyl)aniline, 4-Aminobenzoic acid butyl ester, Butamben, Butambene, Butesin, Butesine, Butoform, Butsein, Butyl 4-aminobenzoate, Butyl Keloform, Butyl p-aminobenzoate, Butylcaine, Hemoride, NSC 128464, Planoform, Scuroform, Scuroforme, n-Butyl 4-aminobenzoate, n-Butyl p-aminobenzoate, p-Aminobenzoic acid butyl ester, p-Aminobenzoic acid n-butyl ester |
| InChI Formula | InChI=1S/C11H15NO2/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7H,2-3,8,12H2,1H3 |
| SMILES | CCCCOC(=O)c1ccc(N)cc1 |
| IUPAC Name | butyl 4-aminobenzoate |
| Molecular Weight (g/mol) | 193.24 |
| Formula Weight | 193.1103 |