Learn More
Butamben, MedChemExpress
Product Code. 16401118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
16401118 5 g 5g
1 options
Product Code. 16401118
Brand: Medchem Express HYB1430
MedChemExpress Butamben (Butyl 4-aminobenzoate) results in long-lasting relief from pain, without impairing motor function or other sensory functions.
Butamben (Butyl 4-aminobenzoate) results in long-lasting relief from pain, without impairing motor function or other sensory functions.
Specifications
Chemical Name or Material Butamben
CAS 94-25-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H15NO2
Quantity 5 g
Synonym Butyl 4-aminobenzoate
Solubility Information DMSO : ≥ 100 mg/mL (517.49 mM) ∣H2O : 0.1 mg/mL (0.52 mM; Need ultrasonic)
SMILES O=C(OCCCC)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 193.24
Formula Weight 193.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation