Learn More
Butamben, LoGiCal
Product Code. 30461888
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30461888
Brand: LoGiCal LGCFOR1802.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Butamben
CAS 94-25-7
Analyte or Component Names Butamben
Shipping Condition Room Temperature
Molecular Formula C11 H15 N O2
Certifications/Compliance ISO 9001
Synonym Butyl Aminobenzoate
InChI Formula InChI=1S/C11H15NO2/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7H,2-3,8,12H2,1H3
Solution Type Neat
SMILES CCCCOC(=O)c1ccc(N)cc1
IUPAC Name butyl 4-aminobenzoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 193.24
Quantity 10 mg
Formula Weight 193.1103 g/mol