Learn More
Uvex Busingess Blue PU 84272 Boot - Width 11
Uvex 1 shoes designed for classic business look; for in the office or on the factory floor
Brand:  Uvex 8427242
Additional Details : Weight : 1.23800kg
 View more versions of this product
Product Code. 17234501
  • 1530.00 NOK / 1 pair
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
Uvex 1 business Desert Boot 8427.2 S3 SRC
  • Safe, smart, stylish
  • Fashionably business look with trendy design
  • Generous fit with wide steel toe cap
  • Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
  • Uvex Climazone technology ensures optimum wearer comfort
  • Breathable, water-resistant high-tech microvelour upper material
  • Textile lining provides optimum breathability
  • Made from synthetic materials, suitable for people allergic to chrome
  • Shock absorbing, durable mono density polyurethane sole unit
  • Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
  • Soft padded collar and tongue
  • Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 8427.2
  • Standard EN ISO 20345:2011 S3 SRC
  • Sizes: 39 to 52
Specifications
Specifications
Blue
42
Steel
EN ISO 20345:2011
Standard width
Yes
S3
Microvelour
Polyurethane (PU)
No
Unisex
Uvex 1
No
SRC