Learn More
Buerkle™ Polyethylene Spigot 'Air'
Product Code. 10214401 Shop All Bürkle Products
Change view
Click to view available options
Material:
Transparent PE spigot, seal made of natural tubber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10214401 Transparent PE spigot, seal made of natural tubber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10214401 Supplier Buerkle™ Supplier No. 05330010
  • ONLINE EXCLUSIVE PRICE
    81.35 NOK
    80.21 NOK / Pack of 1
    Save 1.14 NOK
    1% Off
Estimated Shipment: 02-12-2026
No additional ventilation opening required, PE, transparent, 3/4in.thread connection out-Side.
For use if container ventilation is required. Polyethylene construction, transparent, with an external 3/4 in. thread connection.
  • Self-venting
  • NW 13mm
  • Seal made of natural rubber
TRUSTED_SUSTAINABILITY
Specifications
Closure Type NW 13 mm. 3/4 in. thread connection outside
Material Transparent PE spigot, seal made of natural tubber
Product Type Self-venting Air Spigot