Learn More
Buerkle™ Eyes Fresh Mini Eyewash Station
Product Code. 10465192 Shop All Bürkle Products
Change view
Click to view available options
Mounting:
Faucet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
10465192 Faucet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10465192 Supplier Buerkle™ Supplier No. 25150100
Estimated Shipment: 16-12-2026
Ideal for all work places where aggressive substances are used
  • Allows normal operation of tap
  • Ready for immediate use
  • Can be freely positioned for easy rinsing
  • Rinses both eyes simultaneously
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN 15154-2
For Use With (Application) Standard Cold Water Tap
Material Aluminum
Mounting Faucet
Product Type Eyewash Station