missing translation for 'onlineSavingsMsg'
Learn More

Bupropion, 99.91%, MP Biomedicals™

Product Code. 11493142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
15 mg
Unit Size:
15mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493142 15 mg 15mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493142 Supplier MP Biomedicals Supplier No. 0215376015

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 31677-93-7
Color White
Assay Percent Range 0.9991
Molecular Formula C13H19Cl2NO
Quantity 15 mg
Synonym bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
InChI Key HEYVINCGKDONRU-UHFFFAOYSA-N
SMILES CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
Molecular Weight (g/mol) 276.201
PubChem CID 62884
Formula Weight 276.2
Physical Form Powder
Chemical Name or Material Bupropion
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.