Learn More
Epredia™ Buffered Zinc Formalin Histology Fixative
Enhance nuclear detail in immunohistochemistry techniques with Epredia™ Buffered Zinc Formalin.
Brand: Epredia™ 5705ZF
Additional Details : CAS Number : 7732-18-5 Weight : 18.90000kg
Description
Easy-to-pour bottle helps reduce spills.Specifications
Ea. | |
Formalin | |
5 gallon Cubitainer |