Learn More
Buffer Solution pH9.225 BS formulated, Standard Solution ready to use for pH measurement, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
CAS | 1303-96-4 |
Color | Undesignated |
pH | 9.225 |
Physical Form | Liquid |
Quantity | 1 L |
Packaging | Plastic container |
Product Content Correction
Product Title
Buffer Solution pH9.225 BS formulated, Standard Solution ready to use for pH measurement, Fisher Chemical >