Learn More
Buffer solution, pH 8.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
397.80 NOK - 1161.00 NOK
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366089
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042420.AP
500 mL
397.80 NOK
500mL
View alternative products
Save Up to 
»
In Stock
to see stock.
11304588
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042420.K2
1 L
626.40 NOK
1 L
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 08-07-2024
to see stock.
11314588
FDS missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Alfa Aesar
042420.K7
4 L
1161.00 NOK
4 L
Voir les produits de remplacement
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 15-07-2024
Description
Buffer solution (pH 8) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Vidéos
FDS
Documents
Documents
Certificats
RUO – Research Use Only