missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 6.8, 100 mM KCl, Honeywell Fluka™

Product Code. 15627040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15627040 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15627040 Supplier Honeywell-Fluka Supplier No. 751301L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1336-21-6
pH 6.8
Quantity 1 L
Solution Type Buffer
Molecular Formula H5NO
Linear Formula 3 EtOAc/EtOH
MDL Number MFCD00066650
Packaging Plastic Bottle
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Chemical Name or Material Buffer solution, pH 6.8, 100 mM KCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.