Learn More
Buffer Solution pH 4, Scharlab
Product Code. p-7083571
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Product Code. 11349893
Brand: Scharlab SO10040250
Potassium hydrogen phthalate buffer for use in calibrating pH meters in analytical chemistry.
Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.
Chemical Identifiers
Synonym Potassium hydrogen phthalate buffer
Specifications
Density 1.01g/cm3
pH 4.00
Physical Form Solution
Quantity 250 mL
Formulation Per liter: 10.21g Potassium hydrogen phthalate. Contains preservative.
Traceability to NIST Traceable to NIST
Solubility Information Miscible in water (20°C)