Learn More
Buffer Solution pH 2, Scharlab
Product Code. 11329893
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329893 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11329893 Supplier Scharlab Supplier No. SO10221000
Edge
For use in buffer solutions.
Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.
Specifications
Density ∼1.00g/cm3
pH 2.00
Physical Form Solution
Quantity 1 L
Formulation Per liter: 6.43g Citric acid, 2.40g Sodium hydroxide and 6.13mL Hydrochloric acid
Traceability to NIST Traceable to NIST
Solubility Information Miscible in water (20°C)