missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution for HPCE, 50 mM sodium phosphate, pH 7.0 (25 °C), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
50mL
Specifications
Specifications
| Absorbance | 1: 0 to 0.02; 2: 0 to 0.05; 3: 0 to 0.25 |
| Chemical Name or Material | Buffer solution HPCE pH 7.0 |
| Color | Colorless |
| Concentration | 50 mM |
| pH | 7.0 |
| Physical Form | Liquid |
| Quantity | 50 mL |
| Packaging | Plastic bottle |
| UN Number | NONH for all modes of transport |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction