Learn More
Buffer Solution Duracal pH 7.00 (25°C), Honeywell Fluka™
Product Code. 15632570 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15632570 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15632570 Supplier Honeywell-Fluka Supplier No. 242221500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
500 mL, certified buffer solution traceable to NIST/PTB reference materials, DKD certified
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer Solution Duracal Ph 7.00 (25°C)
Name Note Certified buffer solution traceable to NIST/PTB reference materials, DKD certified
CAS 7778-77-0, 7558-79-4, 7447-40-7
Color Green
Quantity 500 mL
UN Number NONH for all modes of transport