Learn More
Buffer Concentrated Solution pH2.00 (Glycine), NIST Standard for pH measurement, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
Specifications
Specifications
| Color | Colorless |
| pH | 2 |
| Physical Form | Liquid |
| Quantity | 6 Ampules |
| MDL Number | 217672 |
| Packaging | Ampoule |