Learn More
Buffer, 7.00±0.02, Honeywell Fluka™ Hydrion™
Product Code. 15632410 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15632410 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15632410 Supplier Honeywell Fluka Supplier No. 2390891PAK
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
pH 7.00±0.02
  • Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
TRUSTED_SUSTAINABILITY
Tekniske data
Chemical Name or Material Buffer, Hydrion™
pH 7.00 ±0.02
Quantity 1 Pk.
MDL Number MFCD00132752
Packaging Foil envelope
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.