Learn More
Buerkle™ Hose Cutter
Product Code. 10276672 Shop All Bürkle Products
Change view
Click to view available options
For Use With:
Mini Sampler PE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
10276672 Mini Sampler PE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10276672 Supplier Buerkle™ Supplier No. 00069506
  • ONLINE EXCLUSIVE PRICE
    161.00 NOK
    158.06 NOK / Pack of 1
    Save 2.94 NOK
    1% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Edges cut with the hose cutter are always clean and flat, even in the case of thick hoses
Produces cut edges that are always clean and flat, even with thick hoses.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Mini Sampler PE
For Use With (Application) Tri Mming Hoses
Packaging Quantity 1 Pc.
Color Red
Product Type Hose Cutter