Learn More
Bucladesine sodium, MedChemExpress
Product Code. 16414477
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16414477
Brand: Medchem Express HYB0764
MedChemExpress Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.
Bucladesine sodium salt (Dibutyryl-cAMP sodium salt) is a stabilized cyclic AMP (cAMP) analog and a selective PKA activator. Bucladesine sodium salt raises the intracellular levels of cAMP. Bucladesine sodium salt is also a phosphodiesterase (PDE) inhibitor. Bucladesine sodium salt has anti-inflammatory activity and can be used for impaired wound healing.
Specifications
Chemical Name or Material Bucladesine sodium
CAS 16980-89-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H23N5NaO8P
Quantity 50 mg
Synonym Dibutyryl cAMP sodium saltDBcAMP sodium salt
Solubility Information DMSO : 170 mg/mL (345.97 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.76 mM)
SMILES O=C(CCC)O[C@H]1[C@H](N2C(N=CN=C3NC(CCC)=O)=C3N=C2)O[C@@](CO4)([H])[C@@]1([H])OP4(O[Na])=O
Molecular Weight (g/mol) 491.37
Formula Weight 491.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.