Learn More
Micronova™ BucketBinder™ Multi Bucket System
Product Code. 16245152 Shop All Micronova Products
Change view
Click to view available options
Color:
Blue and Red
Blue and Yellow
Blue, Red and Yellow
Unit Size:
Each
Capacity (English):
3 x 6 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size Capacity (English)
16245152 Blue and Red Each -
16255152 Blue and Yellow Each -
16265152 Blue, Red and Yellow Each 3 x 6 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16245152 Supplier Micronova™ Supplier No. BBDBLDBRE
  • ONLINE EXCLUSIVE PRICE
    6950.00 NOK
    6359.62 NOK / Pack of 1
    Save 590.38 NOK
    8% Off
Estimated Shipment: 17-11-2026
Micronova™ BucketBinder™ Multi Bucket Systems are a cost efficient, ergonomic multi bucket system that does away with the need for bulky carts and frames.
The BucketBinder™ kit includes 2 or 3 polypropylene buckets with casters and one or two sets of BucketBinders (bucket connectors). The system offers easier cleaning and storage - in both small and large controlled environments and the sterile core.
  • Polyketone 'hooks' firmly secure two or three rectangular buckets together allowing scale-up or down as required.
  • Easy clip action allows attachment before or after buckets are filled with disinfectant.
  • Sturdy enough to secure two or three buckets - with up to 15 litres of liquid in each
  • Lightweight – less than 2 lbs per pair – BucketBinders™ hold up to strong disinfectants and can be autoclaved and gamma irradiated
  • Once connected, easily house the W-9 W-92 flat wringers, W-9B BoxWringer™ for string mops or W-94 positive pressure flat mop MegaWringer™ wringer
  • Perfectly complements all the Micronova MZone™ FlatMops™, SnapMops™ and SlimLine™ mops.
  • Bucket liners are easily replaced without completely removing BucketBinders™.
  • BucketBinders™ fit the B-7 AC series rectangular buckets.
  • Autoclavable.
Specifications
Product Type Mop Bucket
Capacity (Metric) 44 L (2 Buckets - 22 L/Ea), 12 gal. (2 Buckets - 6 gal./Ea)
Material Polypropylene, Polyketone, Stainless Steel
Autoclavable Autoclavable
Color Blue and Red
For Use With (Application) Cleanroom Surface Cleaning and Disinfecting
For Use With (Equipment) W-9 Series Wringer
Height (Metric) 37 cm
Length (Metric) 55 cm
Quantity 1 Ea.
Sterility Non-sterile
Width (Metric) 65 cm