Learn More
Hettich Lab Technology™ Bucket
Click to view available options
Capacity:
2 x 25 mL
4 x 15 mL
Unit Size:
Each
Description
- Available in a variety of configurations
- Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
Specifications
| Capacity | 4 x 15 mL |
| Bore Size (Metric) | 17.6 OD x 74 mm L |
| Container Type | Bucket |
| No. of Places | 4 |
| Tube Size | 15 mL |