Learn More
Hettich Lab Technology™ Bucket
Product Code. 10220052
Click to view available options
Capacity:
2 x 25 mL
4 x 15 mL
Unit Size:
Each
Product Code. 10220052
Brand: Hettich Lab Technology™ 1369
Bucket comes in a variety of designs to accommodate customized centrifuge loads.
  • Available in a variety of configurations
  • Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
Capacity 4 x 15 mL
Bore Size (Metric) 17.6 OD x 74 mm L
Container Type Bucket
No. of Places 4
Tube Size 15 mL