missing translation for 'onlineSavingsMsg'
Learn More

BTZ043 Racemate, MedChemExpress

Product Code. p-200101515
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471917

Brand: Medchem Express HY13579A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BTZ043 Racemate (BTZ10526038) is the racemate of BTZ043. BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate.

BTZ043 Racemate (BTZ10526038) is the racemate of BTZ043. BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate.

Specifications

Chemical Name or Material BTZ043 Racemate
Physical Form Solid
CAS 957217-65-1
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H16F3N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym BTZ10526038 Benzothiazinone 10526038
Solubility Information DMSO : 25 mg/mL (57.95 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1N=C(N(CC2)CCC32OCC(C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14
Molecular Weight (g/mol) 431.39
Formula Weight 431.39
Percent Purity 98.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.