Learn More
BTZ043 Racemate, MedChemExpress
Product Code. 16471917
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471917 50 mg 50mg
16425768 10 mg 10mg
16408447 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16471917 Supplier Medchem Express Supplier No. HY13579A
MedChemExpress BTZ043 Racemate (BTZ10526038) is the racemate of BTZ043. BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate.
BTZ043 Racemate (BTZ10526038) is the racemate of BTZ043. BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate.
Specifications
Chemical Name or Material BTZ043 Racemate
Physical Form Solid
CAS 957217-65-1
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H16F3N3O5S
For Use With (Application) COVID-19-immunoregulation
Synonym BTZ10526038 Benzothiazinone 10526038
Solubility Information DMSO : 25 mg/mL (57.95 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1N=C(N(CC2)CCC32OCC(C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14
Molecular Weight (g/mol) 431.39
Formula Weight 431.39
Percent Purity 98.77%
Grade Research