missing translation for 'onlineSavingsMsg'
Learn More

BTNL8 Antibody (2187B) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30171439 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171439 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171439 Supplier Novus Biologicals Supplier No. FAB9359JF635
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTNL8 Monoclonal antibody specifically detects BTNL8 in Human samples. It is validated for Flow Cytometry,Western Blot,Simple Western
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTNL8
Applications Flow Cytometry, Western Blot, Simple Western
Classification Monoclonal
Clone 2187B
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias butyrophilin-like 8, butyrophilin-like protein 8, UNQ702/PRO1347
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK-293-derived Human BTNL8, Gln18-Lys238, Accession # Q6UX41-1
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79908
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.