missing translation for 'onlineSavingsMsg'
Learn More

BTG2 Antibody, Novus Biologicals™

Product Code. p-200040143 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25ul
Unit Size:
0.10mL
25µL
Product Code. Quantity unitSize
18429230 25ul 25µL
18790764 - 0.10mL
2 options
This item is not returnable. View return policy

Product Code. 18429230

Brand: Novus Biologicals NBP18121525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

BTG2 Polyclonal specifically detects BTG2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTG2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21
Gene Symbols BTG2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:QEALTEHYKHHWFPEKPSKGSGYRCIRINHKMDPIISRVASQIGLSQPQLHQLLPSELTLWVDPYEVSYRIGEDGSICVLYEEAPLAASCGLLTCKNQVLLG
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7832
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.