missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14879-1-AP-150UL
This item is not returnable.
View return policy
Description
BTG1 is a member of an anti-proliferative gene family that regulates cell growth and differentiation. Expression of this gene is highest in the G0/G1 phases of the cell cycle and downregulated when cells progressed through G1. The encoded protein interacts with several nuclear receptors, and functions as a coactivator of cell differentiation. This locus has been shown to be involved in a t(8; 12)(q24; q22) chromosomal translocation in a case of B-cell chronic lymphocytic leukemia.Specifications
| BTG1 | |
| Polyclonal | |
| Unconjugated | |
| BTG1 | |
| BTG1, Protein BTG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12226, 29618, 694 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62324, P62325, Q63073 | |
| BTG1 | |
| BTG1 Fusion Protein Ag6651 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction