Learn More
Corning™ BTD Valve Head Complete
Product Code. 16802100 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
25 to 100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
16802100 2.5 to 10 mL Each
16812100 25 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16802100 Supplier Corning™ Supplier No. 6851
  • ONLINE EXCLUSIVE PRICE
    1360.00 NOK
    1267.29 NOK / Pack of 1
    Save 92.71 NOK
    6% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
Corning™ valve head complete
TRUSTED_SUSTAINABILITY
Specifications
Description BTD valve head complete
Volume (Metric) 2.5 to 10 mL
For Use With (Application) Dispensing Organic or Inorganic Liquids