Learn More
Gibco™ BSA Fraction V IgG Free Fatty Acid Poor (custom product)
Description
This is a GIBCO Custom Product. Please complete and submit the Custom Media Inquiry Form and a Customer Service Representative will contact you regarding pricing and availability.
Specifications
Specifications
| Content And Storage | 2 to 8°C |
| Description | BSA Fraction V IgG Free Fatty Acid Poor, 100g, Solid, Used for Cell Viability, Proliferation and Function, Mammalian Cell Culture, For Research Use Only, Not for Use in Diagnostic Procedures |
| Form | Solid |
| Product Type | BSA |
| For Use With (Application) | Cell viability, Proliferation and Function, Mammalian Cell Culture |
| Quantity | 100 g |
| Regulatory Status | For research use only |