Learn More
Brannan™ BS 718 Model Soda Glass M50 SP Petroleum Density Hydrometers
Click to view available options
Specific Gravity Range:
0.650 to 0.700 g/mL
0.700 to 0.750 g/mL
0.750 to 0.800 g/mL
0.800 to 0.850 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
1.050 to 1.100 g/mL
Unit Size:
Each
Description
For surface tension 0.600 to 0.650g/mL Shot loaded, length 270mm, subdivisions 0.001g/mL, accuracy ±0.0006g/mL. Calibrated at 15°C.
Specifications
Specifications
| Quantity | 1Pack |
| Type | M50 SP Petroleum Density Hydrometer |
| Accuracy | 0.001g/mL |
| Increments | 0.001 g/mL |
| Certifications/Compliance | ISO 387, ISO 649 |
| Length (Metric) | 270 mm |
| Ballast Type | Steel Shot |
| For Use With (Application) | Petroleum industry and corresponds to IP 160 |
| Specific Gravity Range | 0.750 to 0.800 g/mL |