Learn More
Brannan™ BS 718 : 1991 Density Hydrometer L50
Click to view available options
Specific Gravity Range:
0.75 to 0.80 g/mL
0.80 to 0.85 g/mL
For Use With (Application):
For low surface tension
Unit Size:
Each
Description
- BS 718 : 1991 is generally aligned with ISO 387 and ISO 649
- Calibrated using the accepted units of grams per milliliter (g/mL)
- Available for numerous specific gravity ranges
- Available for both low and medium surface tension
- Includes protective plastic tube
- 330mm in length
Specifications
Specifications
| Type | Density Hydrometer |
| Accuracy | 0.0005 g/mL |
| Increments | 0.0005 g/mL |
| Certifications/Compliance | ISO 387 and ISO 649 |
| Length (Metric) | 330 mm |
| Includes | Protective plastic tube (47 g) |
| Diameter (Metric) Outside | 23 to 27 mm (Bulb) |
| Material | Soda glass, Steel shot ballast, Synthetic wax resin |
| Shipping Weight (Metric) | 103 g |
| Weight (Metric) | 56 g |
| Show More |