Learn More
Emma Safety Footwear Bryce XD Safety Shoes
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
1515.80 NOK
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15948361
|
Emma Safety Footwear
MM33186839 |
39 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15958361
|
Emma Safety Footwear
MM33186840 |
40 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15968361
|
Emma Safety Footwear
MM33186841 |
41 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15978361
|
Emma Safety Footwear
MM33186842 |
42 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15988361
|
Emma Safety Footwear
MM33186843 |
43 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15998361
|
Emma Safety Footwear
MM33186844 |
44 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15908371
|
Emma Safety Footwear
MM33186845 |
45 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15918371
|
Emma Safety Footwear
MM33186846 |
46 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15928371
|
Emma Safety Footwear
MM33186847 |
47 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15938371
|
Emma Safety Footwear
MM33186848 |
48 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15948371
|
Emma Safety Footwear
MM33186849 |
49 |
1515.80 NOK
1 pair |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Microfiber construction
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver
Specifications
Bryce XD | |
Microfiber |