Learn More
Emma Safety Footwear Bryce Safety Shoes
Product Code. 15998351
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998351 45 1 pair
15918351 37 1 pair
15928351 38 1 pair
15938351 39 1 pair
15948351 40 1 pair
15958351 41 1 pair
15968351 42 1 pair
15978351 43 1 pair
15988351 44 1 pair
15908361 46 1 pair
15918361 47 1 pair
15928361 48 1 pair
15938361 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15998351 Supplier Emma Safety Footwear Supplier No. MM33084845
  • ONLINE EXCLUSIVE PRICE
    1794.79 NOK
    981.75 NOK / 1 pairs
    Save 813.04 NOK
    45% Off
Estimated Shipment: 30-03-2027
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Microfiber construction
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Bryce
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 45
Toe Material Steel