Learn More
BRP16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20169-1-AP-20UL
 View more versions of this product
Product Code. 16859674
  • 1710.33 NOK / 20µL
Description
Hgh1 is a protein coding gene.
Specifications
Specifications
BRP16
Polyclonal
Unconjugated
HGH1
Brain protein 16, BRP16, C8orf30A
Rabbit
Antigen Affinity Chromatography
RUO
51236, 59053
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8C3I8, Q9BTY7
HGH1
BRP16 Fusion Protein Ag13998
20 μL
Primary
Human, Mouse