Learn More
Steris™ Browne™ Control™ Autoclave Indicator Valves
Product Code. 12725335
Change view
Click to view available options
Color:
Green Spot
White Spot
Yellow Spot
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12725335 Green Spot Pack of 100
12715335 Yellow Spot Pack of 100
12745335 White Spot Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12725335 Supplier Steris™ Supplier No. 7303AB
Add to basket
Add to basket
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
  • The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
  • Various colors
  • Glass tubes
TRUSTED_SUSTAINABILITY
Specifications
Color Green Spot
For Use With (Application) For monitoring porous load steam sterilization processes
For Use With (Equipment) Autoclave
Product Type Browne Control Tube
Temperature (Metric) 160°C
Quantity 100 Pk.
Sterilization Indicator Yes
Sterilization Time 60 min.
Sterilization Method Steam, Dry Heat