Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
Product Code. 10215443
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Tolerance:
±0.02 mL
±0.025 mL
±0.03 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Standard Taper Joint Tolerance unitSize
10215443 25 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
12688665 100 mL 12/21 NS Standard Ground ±0.1 mL Pack of 2
10598472 1000 mL 24/29 NS Standard Ground ±0.4 mL Each
11736712 100 mL 14/23 NS Standard Ground ±0.1 mL Pack of 2
13177939 10 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11796702 10 mL 7/16 NS Standard Ground ±0.025 mL Pack of 2
10761782 2000 mL 29/32 NS Standard Ground ±0.6 mL Each
10139342 20 mL 10/19 NS Standard Ground ±0.03 mL Pack of 2
11706712 20 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11716722 200 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11726722 250 mL 14/23 NS Standard Ground ±0.15 mL Pack of 2
11652229 25 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
11736722 500 mL 19/26 NS Standard Ground ±0.25 mL Pack of 2
11726712 50 mL 12/21 NS Standard Ground ±0.06 mL Pack of 2
11786702 5 mL 7/16 NS Standard Ground ±0.025 mL Pack of 2
11752303 50 mL 14/23 NS Standard Ground ±0.08 mL Pack of 2
11714116 5 mL 10/19 NS Standard Ground ±0.04 mL Pack of 2
10199192 5 mL 10/19 NS Standard Ground ±0.02 mL Pack of 2
18 options
Product Code. 10215443
Brand: Hirschmann™ 2640171
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Color Brown
Capacity (Metric) 25 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 110 mm
Limit of Error ±0.06 mL
Standard Taper Joint 12/21 NS Standard Ground
Tolerance ±0.06 mL
Graduated Yes
Class A