Learn More
Hirschmann™ Class A Volumetric Flasks, Amber Glass
Product Code. 30090420
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
150 mL
200 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Standard Taper Joint unitSize
30090420 150 mL 14/23 NS Standard Ground Pack of 2
11859050 100 mL 12/21 NS Standard Ground Pack of 2
10116590 1000 mL 24/29 NS Standard Ground Each
11786722 100 mL 14/23 NS Standard Ground Pack of 2
11878891 10 mL 10/19 NS Standard Ground Pack of 2
10761041 2000 mL 29/32 NS Standard Ground Each
11756722 10 mL 7/16 NS Standard Ground Pack of 2
11766722 20 mL 10/19 NS Standard Ground Pack of 2
11776722 25 mL 10/19 NS Standard Ground Pack of 2
11716742 200 mL 14/23 NS Standard Ground Pack of 2
11682229 20 mL 12/21 NS Standard Ground Pack of 2
11736742 500 mL 19/26 NS Standard Ground Pack of 2
11706742 50 mL 12/21 NS Standard Ground Pack of 2
11839600 50 mL 14/23 NS Standard Ground Pack of 2
13187939 5 mL 10/19 NS Standard Ground Pack of 2
15 options
Product Code. 30090420
Brand: Hirschmann™ 2640283
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks, Amber Glass are manufactured from durable DURAN™ glass tubing.
  • With NS ground and hollow amber glass stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Calibration To Contain
Certifications/Compliance DIN EN ISO 1042
Closure Color Brown
Closure Material Polyethylene
Color Brown
Capacity (Metric) 150 mL
Material Borosilicate Glass (3.3)
Height (Metric) 180 ± 5 mm
Includes Polyethylene Stopper
Limit of Error ± 0.1 mL
No. per Pack 2/Pack
Standard Taper Joint 14/23 NS Standard Ground
Tolerance ± 0.15 mL
Writing Surface White enamel color
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Sterility Non-sterile
Class A
Bottom Shape Round
Closure Size NS 14/23